Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 2-methylpropane-2-sulfinate 98%

Supplier: AMBEED, INC
Warning

A1070533-100MG A1070533-1G A1070533-250MG
76671-860EA 84 USD InStock
76671-860 76671-862 76671-864
Sodium 2-methylpropane-2-sulfinate 98%
Sodium 2-methylpropane-2-sulfinate
Precaución: for Research Use Only


Formula: C₄H₉NaO₂S
MDL Number: MFCD28166511
CAS Number: 69152-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty