Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Phenyl-1,3-dioxolan-2-one 95%

Supplier: AMBEED, INC
Warning

A1070595-100MG A1070595-1G
76671-866EA 81 USD InStock
76671-866 76671-868
4-Phenyl-1,3-dioxolan-2-one 95%
4-Phenyl-1,3-dioxolan-2-one
Precaución: for Research Use Only


Formula: C₉H₈O₃
MDL Number: MFCD20134160
CAS Number: 4427-92-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty