Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-(3-aminobenzamido)benzoate 95%

Supplier: AMBEED, INC
Warning

A1070722-100MG A1070722-250MG A1070722-1G
76671-870EA 36 USD InStock
76671-870 76671-872 77619-780
Ethyl 3-(3-aminobenzamido)benzoate 95%
Ethyl 3-(3-aminobenzamido)benzoate
Precaución: for Research Use Only


Formula: C₁₆H₁₆N₂O₃
MDL Number: MFCD28714749
CAS Number: 1307391-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty