Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Disodium (ethyl phosphono)formate 95%

Supplier: AMBEED, INC
Warning

A1072908-100MG A1072908-1G A1072908-250MG A1072908-5G
76671-924EA 24 USD InStock
76671-924 76671-926 76671-928 77619-794
Disodium (ethyl phosphono)formate 95%
Disodium (ethyl phosphono)formate
Precaución: for Research Use Only


Formula: C₃H₅Na₂O₅P
MDL Number: MFCD25977182
CAS Number: 55920-24-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty