Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-(2-Iodophenyl)-3-methyl-1,2,4-oxadiazole 95%

Supplier: AMBEED, INC
Warning

A1075482-100MG A1075482-250MG
76671-948EA 69 USD InStock
76671-948 76671-950
5-(2-Iodophenyl)-3-methyl-1,2,4-oxadiazole 95%
5-(2-Iodophenyl)-3-methyl-1,2,4-oxadiazole
Precaución: for Research Use Only


Formula: C₉H₇IN₂O
MDL Number: MFCD21091817
CAS Number: 1283108-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty