Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetonitrile 98%

Supplier: AMBEED, INC
Warning

A1079843-100MG A1079843-250MG
76672-016EA 104 USD InStock
76672-016 76672-018
2-(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetonitrile 98%
2-(2,4-Dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)acetonitrile
Precaución: for Research Use Only


Formula: C₆H₅N₃O₂
MW: 151.1228 g/mol
MDL Number: MFCD10692117
CAS Number: 130278-40-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty