Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6-Trihydroxybenzene-1,3-dicarbaldehyde 95%

Supplier: AMBEED, INC
Danger

A1082348-250MG A1082348-50MG A1082348-100MG
76672-058EA 105 USD InStock
76672-058 76672-060 77619-806
4,5,6-Trihydroxybenzene-1,3-dicarbaldehyde 95%
4,5,6-Trihydroxybenzene-1,3-dicarbaldehyde
Precaución: for Research Use Only


Formula: C₈H₆O₅
MDL Number: MFCD24676794
CAS Number: 58343-12-7
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty