Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-3-chloro-2-methylbenzoic acid 95%

Supplier: AMBEED, INC
Warning

A1082878-100MG A1082878-5G
76672-066EA 37 USD InStock
76672-066 76672-068
5-Bromo-3-chloro-2-methylbenzoic acid 95%
5-Bromo-3-chloro-2-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₆BrClO₂
MDL Number: MFCD24108594
CAS Number: 1540188-38-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty