Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Propan-2-yl)-1H-pyrazole-3-sulfonamide 95%

Supplier: AMBEED, INC
Warning

A1087728-100MG A1087728-250MG
76672-120EA 80 USD InStock
76672-120 77619-818
1-(Propan-2-yl)-1H-pyrazole-3-sulfonamide 95%
1-(Propan-2-yl)-1H-pyrazole-3-sulfonamide
Precaución: for Research Use Only


Formula: C₆H₁₁N₃O₂S
MDL Number: MFCD30329853
CAS Number: 1696838-10-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty