Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-3-methanesulfonylbenzene 95%

Supplier: AMBEED, INC
Warning

A1094330-1G A1094330-250MG
76672-178EA 142 USD InStock
76672-178 76672-180
1-Ethynyl-3-methanesulfonylbenzene 95%
1-Ethynyl-3-methanesulfonylbenzene
Precaución: for Research Use Only


Formula: C₉H₈O₂S
MDL Number: MFCD13189809
CAS Number: 573982-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty