Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-(tetrahydropyran-4-yl)pyridine 95%

Supplier: AMBEED, INC
Danger

A1096612-1G A1096612-250MG A1096612-50MG
76672-212EA 650 USD InStock
76672-212 76672-214 76672-216
2-Bromo-5-(tetrahydropyran-4-yl)pyridine 95%
2-Bromo-5-(tetrahydropyran-4-yl)pyridine
Precaución: for Research Use Only


Formula: C₁₀H₁₂BrNO
MDL Number: MFCD12033507
CAS Number: 1159817-81-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty