Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Iodo-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 95%

Supplier: AMBEED, INC
Warning

A1103545-100MG A1103545-1G A1103545-250MG
76672-250EA 55 USD InStock
76672-250 76672-252 76672-254
5-Iodo-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 95%
5-Iodo-1-methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine
Precaución: for Research Use Only


Formula: C₅H₅F₃IN₃
CAS Number: 2092046-65-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty