Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methyl-5-(morpholin-4-yl)pyridin-2-amine 95%

Supplier: AMBEED, INC
Warning

A1104684-100MG A1104684-1G A1104684-250MG
76672-272EA 69 USD InStock
76672-272 76672-274 76672-276
3-Methyl-5-(morpholin-4-yl)pyridin-2-amine 95%
3-Methyl-5-(morpholin-4-yl)pyridin-2-amine
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃O
MW: 193.2456 g/mol
MDL Number: MFCD26735760
CAS Number: 1554988-88-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty