Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluorobenzyl mercaptan 95%

Supplier: AMBEED, INC
Danger

A1109416-1G A1109416-250MG A1109416-5G
76672-362EA 50 USD InStock
76672-362 76672-364 76672-366
2-Fluorobenzyl mercaptan 95%
2-Fluorobenzyl mercaptan
Precaución: for Research Use Only


Formula: C₇H₇FS
MDL Number: MFCD00041467
CAS Number: 72364-46-6
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty