Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diphenylpropan-1-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1110103-100MG A1110103-1G A1110103-250MG
76672-376EA 26 USD InStock
76672-376 76672-378 76672-380
1,3-Diphenylpropan-1-amine hydrochloride 95%
1,3-Diphenylpropan-1-amine hydrochloride
Precaución: for Research Use Only


Formula: C₁₅H₁₈ClN
MDL Number: MFCD12197109
CAS Number: 5454-35-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty