Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclohexanesulfinic acid sodium salt 95%

Supplier: AMBEED, INC
Warning

A1125877-100MG A1125877-250MG
76672-602EA 35 USD InStock
76672-602 76672-604
Cyclohexanesulfinic acid sodium salt 95%
Cyclohexanesulfinic acid sodium salt
Precaución: for Research Use Only


Formula: C₆H₁₁NaO₂S
MW: 170.2051 g/mol
MDL Number: MFCD20233346
CAS Number: 74829-95-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty