Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 3,3,3-trifluoropropane-1-sulfinate 98+%

Supplier: AMBEED, INC
Warning

A1126031-100MG A1126031-1G A1126031-250MG A1126031-5G
76672-618EA 38 USD InStock
76672-618 76672-620 76672-622 76672-624
Sodium 3,3,3-trifluoropropane-1-sulfinate 98+%
Sodium 3,3,3-trifluoropropane-1-sulfinate
Precaución: for Research Use Only


Formula: C₃H₄F₃NaO₂S
MDL Number: MFCD18379709
CAS Number: 1263377-91-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty