Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Bromomethyl)benzene-1-sulfonyl fluoride 95%

Supplier: AMBEED, INC
Danger

A1127175-1G A1127175-250MG
76672-646EA 650 USD InStock
76672-646 76672-648
3-(Bromomethyl)benzene-1-sulfonyl fluoride 95%
3-(Bromomethyl)benzene-1-sulfonyl fluoride
Precaución: For research use only.


Formula: C₇H₆BrFO₂S
MW: 253.0887 g/mol
MDL Number: MFCD31445396
CAS Number: 79686-36-5
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty