Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethynyl-2,4,5-trifluorobenzene 95%

Supplier: AMBEED, INC
Danger

A1133049-100MG A1133049-1G
76672-770EA 63 USD InStock
76672-770 76672-772
1-Ethynyl-2,4,5-trifluorobenzene 95%
1-Ethynyl-2,4,5-trifluorobenzene
Precaución: for Research Use Only


Formula: C₈H₃F₃
MDL Number: MFCD11655774
CAS Number: 1097874-78-6
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty