Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-hydroxy-3-phenylpropanoate 98%

Supplier: AMBEED, INC
Warning

A1135776-1G A1135776-250MG
76672-792EA 451 USD InStock
76672-792 76672-794
Ethyl 2-hydroxy-3-phenylpropanoate 98%
Ethyl 2-hydroxy-3-phenylpropanoate
Precaución: for Research Use Only


Formula: C₁₁H₁₄O₃
MW: 194.2271 g/mol
MDL Number: MFCD00128177
CAS Number: 15399-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty