Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Chloro-2-(methoxymethyl)-6-methylpyrimidine 95%

Supplier: AMBEED, INC
Warning

A1137952-100MG A1137952-250MG
76672-812EA 38 USD InStock
76672-812 76672-814
4-Chloro-2-(methoxymethyl)-6-methylpyrimidine 95%
4-Chloro-2-(methoxymethyl)-6-methylpyrimidine
Precaución: for Research Use Only


Formula: C₇H₉ClN₂O
MDL Number: MFCD11043806
CAS Number: 3122-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty