Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

BIBF-0775 97%

Supplier: AMBEED, INC
Warning

A1142642-10MG A1142642-1MG A1142642-50MG A1142642-5MG
76672-902EA 63 USD InStock
76672-902 76672-904 76672-906 76672-908
BIBF-0775 97%
BIBF-0775
Precaución: for Research Use Only


Formula: C₃₁H₃₄N₃₄O₂
MW: 494.64 g/mol
MDL Number: MFCD25976645
CAS Number: 334951-90-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty