Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PZ-2891 98%

Supplier: AMBEED, INC
Warning

A1142882-10MG A1142882-1MG A1142882-50MG A1142882-5MG
76672-948EA 61 USD InStock
76672-948 76672-950 76672-952 76672-954
PZ-2891 98%
PZ-2891
Precaución: for Research Use Only


Formula: C₂₀H₂₃N₅O
MDL Number: MFCD31812779
CAS Number: 2170608-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty