Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ir[dF(Me)ppy]2(dtbbpy)PF6 97%

Supplier: AMBEED, INC
Warning

A1143090-100MG A1143090-1G A1143090-250MG A1143090-50MG
76672-990EA 28 USD InStock
76672-990 76672-992 76672-994 76672-996
Ir[dF(Me)ppy]2(dtbbpy)PF6 97%
Ir[dF(Me)ppy]2(dtbbpy)PF6
Precaución: for Research Use Only


Formula: C₄₂H₄₀F₁₀IrN₄P
MDL Number: MFCD31619196
CAS Number: 1335047-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty