Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,6-Dichloro-1,2,4-triazine 97%

Supplier: AMBEED, INC
Danger

A1143455-100MG A1143455-1G
76673-066EA 57 USD InStock
76673-066 76673-068
3,6-Dichloro-1,2,4-triazine 97%
3,6-Dichloro-1,2,4-triazine
Precaución: for Research Use Only


Formula: C₃HCl₂N₃
MDL Number: MFCD22548154
CAS Number: 99584-48-2
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty