Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride 97%

Supplier: AMBEED, INC
Warning

A1143630-100MG A1143630-1G A1143630-250MG A1143630-5G
76673-082EA 13 USD InStock
76673-082 76673-084 76673-086 76673-088
(R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride 97%
(R)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride
Precaución: for Research Use Only


Formula: C₁₇H₂₇ClN₂O
MDL Number: MFCD22572716
CAS Number: 112773-90-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty