Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate] 99%

Supplier: AMBEED, INC
Warning

A1144500-100G A1144500-25G
76673-164EA 15 USD InStock
76673-164 76673-166
Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate] 99%
Trimethylolpropane tris[3-(2-methylaziridin-1-yl)propionate]
Precaución: for Research Use Only


Formula: C₂₄H₄₁N₃O₆
MW: 467.61 g/mol
MDL Number: MFCD00192542
CAS Number: 64265-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty