Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N2,N2'-Dimethyl-[1,1'-binaphthalene]-2,2'-diamine 97%

Supplier: AMBEED, INC
Warning

A1144624-1G A1144624-250MG
76673-174EA 89 USD InStock
76673-174 76673-176
N2,N2'-Dimethyl-[1,1'-binaphthalene]-2,2'-diamine 97%
N2,N2'-Dimethyl-[1,1'-binaphthalene]-2,2'-diamine
Precaución: for Research Use Only


Formula: C₂₂H₂₀N₂
MDL Number: MFCD10566977
CAS Number: 93621-64-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty