Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A1145052-100MG A1145052-1G A1145052-250MG A1145052-5G
76673-198EA 17 USD InStock
76673-198 76673-200 76673-202 76673-204
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester 97%
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₃BN₂O₂
MW: 262.16 g/mol
MDL Number: MFCD13176540
CAS Number: 1233526-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty