Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Tetrahydropyran-2-yl)-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1H-indazole 95%

Supplier: AMBEED, INC
Warning

A1145094-100MG A1145094-1G A1145094-250MG
76673-240EA 28 USD InStock
76673-240 76673-242 76673-244
1-(Tetrahydropyran-2-yl)-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1H-indazole 95%
1-(Tetrahydropyran-2-yl)-4-(4,4,5,5-tetramethyl[1,3,2]dioxaborolan-2-yl)-1H-indazole
Precaución: for Research Use Only


Formula: C₁₈H₂₅BN₂O₃
MDL Number: MFCD18207194
CAS Number: 956388-05-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty