Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1,4-Dioxan-2-yl)methanamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A1145184-10G A1145184-1G A1145184-250MG
76673-256EA 857 USD InStock
76673-256 76673-258 76673-260
(1,4-Dioxan-2-yl)methanamine hydrochloride 95%
(1,4-Dioxan-2-yl)methanamine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO₂
MDL Number: MFCD06739050
CAS Number: 1185156-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty