Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,4S)-Dimethyl 1-((R)-1-(phenylamino)-2-(p-tolyl)ethyl)-7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate 98%

Supplier: AMBEED, INC
Warning

A1145658-25MG A1145658-50MG
76673-304EA 59.42 USD InStock
76673-304 76673-306 76673-308 76673-310 76673-312
(1R,4S)-Dimethyl 1-((R)-1-(phenylamino)-2-(p-tolyl)ethyl)-7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate 98%
(1R,4S)-Dimethyl 1-((R)-1-(phenylamino)-2-(p-tolyl)ethyl)-7-oxabicyclo[2.2.1]hepta-2,5-diene-2,3-dicarboxylate
Precaución: for Research Use Only


Formula: C₂₅H₂₅NO₅
MW: 419.47 g/mol
MDL Number: MFCD31807643
CAS Number: 1534358-79-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty