Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(2-(1H-1,2,4-Triazol-1-yl)-5-(trifluoromethoxy)phenyl)-4-(cyclopropylmethoxy)-3-methoxybenzamide 98%

Supplier: AMBEED, INC
Warning

A1145736-10MG A1145736-1MG A1145736-50MG A1145736-5MG
76673-364EA 45 USD InStock
76673-364 76673-366 76673-368 76673-370
N-(2-(1H-1,2,4-Triazol-1-yl)-5-(trifluoromethoxy)phenyl)-4-(cyclopropylmethoxy)-3-methoxybenzamide 98%
N-(2-(1H-1,2,4-Triazol-1-yl)-5-(trifluoromethoxy)phenyl)-4-(cyclopropylmethoxy)-3-methoxybenzamide
Precaución: for Research Use Only


Formula: C₂₁H₁₉F₃N₄O₄
MW: 448.4 g/mol
MDL Number: MFCD31813758
CAS Number: 2313526-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty