Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Uridine 5'-(trihydrogen diphosphate) P'-(2-deoxy-2-fluoro-α-D-glucopyranosyl) ester 97%

Supplier: AMBEED, INC
Warning

A1146308-100MG A1146308-25MG A1146308-50MG
76673-410EA 1599 USD InStock
76673-410 76673-412 76673-414
Uridine 5'-(trihydrogen diphosphate) P'-(2-deoxy-2-fluoro-α-D-glucopyranosyl) ester 97%
Uridine 5'-(trihydrogen diphosphate) P'-(2-deoxy-2-fluoro-α-D-glucopyranosyl) ester
Precaución: for Research Use Only


Formula: C₁₅H₂₃FN₂O₁₆P₂
CAS Number: 67341-43-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty