Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Copper(II) pyrophosphate hydrate 99%

Supplier: AMBEED, INC
Warning

A1147166-100G A1147166-25G A1147166-500G
76673-510EA 8 USD InStock
76673-510 76673-512 76673-514
Copper(II) pyrophosphate hydrate 99%
Copper(II) pyrophosphate hydrate
Precaución: for Research Use Only


Formula: Cu₂P₂O₇·xH₂O
MW: 301.02 g/mol (anhy)
MDL Number: MFCD00150225
CAS Number: 16570-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty