Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(11bR)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide 98%

Supplier: AMBEED, INC
Warning

A1147887-100MG A1147887-1G A1147887-250MG A1147887-5G
76673-630EA 113 USD InStock
76673-630 76673-632 76673-634 76673-636
(11bR)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide 98%
(11bR)-8,9,10,11,12,13,14,15-Octahydro-4-hydroxy-2,6-bis(2,4,6-tricyclohexylphenyl)-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin 4-oxide
Precaución: for Research Use Only


Formula: C₆₈H₈₉O₄P
MW: 1001.4000 g/mol
CAS Number: 1676072-26-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty