Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(2E,4E)-5-Phenyl-2,4-pentadienoic acid 99+%

Supplier: AMBEED, INC
Warning

A1148989-1G A1148989-5G A1148989-25G
76673-816EA 8 USD InStock
76673-816 76673-818 77620-066
(2E,4E)-5-Phenyl-2,4-pentadienoic acid 99+%
(2E,4E)-5-Phenyl-2,4-pentadienoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₀O₂
MDL Number: MFCD00014018
CAS Number: 28010-12-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty