Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-N-(2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine hydrochloride 98%

Supplier: AMBEED, INC
Danger

A1149007-100G A1149007-1G A1149007-25G A1149007-5G
76673-828EA 28 USD InStock
76673-828 76673-830 76673-832 76673-834
(Z)-N-(2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine hydrochloride 98%
(Z)-N-(2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₂₃ClN₄O₃S
MW: 350.8647 g/mol
MDL Number: MFCD00069339
CAS Number: 358638-36-5
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty