Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-(3-Chloroprop-1-en-1-yl)benzene 95% stabilized

Supplier: AMBEED, INC
Danger

Stabilization: Stabilised with sodium carbonate (Na₂CO₃)

A1149049-25G A1149049-5G
76673-840EA 16 USD InStock
76673-840 76673-842
(E)-(3-Chloroprop-1-en-1-yl)benzene 95% stabilized
(E)-(3-Chloroprop-1-en-1-yl)benzene
Precaución: for Research Use Only


Formula: C₉H₉Cl
MDL Number: MFCD00000986
CAS Number: 21087-29-6
UN: 2922
ADR: 8,6.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty