Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethyl-1-methyl-1H-imidazol-3-ium sodium sulfate 98%

Supplier: AMBEED, INC
Danger

A1149071-1G A1149071-25G A1149071-5G
76673-844EA 10 USD InStock
76673-844 76673-846 76673-848
3-Ethyl-1-methyl-1H-imidazol-3-ium sodium sulfate 98%
3-Ethyl-1-methyl-1H-imidazol-3-ium sodium sulfate
Precaución: for Research Use Only


Formula: C₆H₁₁N₂NaO₄S
CAS Number: 674282-83-8
UN: 1760
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty