Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-2-oxoethyl)-1-methyl-2-(3-(trifluoromethyl)phenyl)-1H-benzo[d]imidazol-3-ium iodide 98%

Supplier: AMBEED, INC
Warning

A1149112-10MG A1149112-1MG A1149112-25MG A1149112-50MG A1149112-5MG
76673-868EA 65 USD InStock
76673-868 76673-870 76673-872 76673-874 76673-876
3-(2-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-2-oxoethyl)-1-methyl-2-(3-(trifluoromethyl)phenyl)-1H-benzo[d]imidazol-3-ium iodide 98%
3-(2-(((1R,2S,5R)-2-Isopropyl-5-methylcyclohexyl)oxy)-2-oxoethyl)-1-methyl-2-(3-(trifluoromethyl)phenyl)-1H-benzo[d]imidazol-3-ium iodide
Precaución: for Research Use Only


Formula: C₂₇H₃₂F₃IN₂O₂
MDL Number: MFCD31813866
CAS Number: 2101317-21-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty