Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(((2-((3-Methoxybenzyl)carbamoyl)-4-oxo-1,4-dihydrothieno[2,3-d]pyrimidin-5-yl)methoxy)methyl)benzoic acid 99%

Supplier: AMBEED, INC
Warning

A1149115-10MG A1149115-1MG A1149115-25MG A1149115-5MG
76673-896EA 95 USD InStock
76673-896 76673-898 76673-900 76673-902
4-(((2-((3-Methoxybenzyl)carbamoyl)-4-oxo-1,4-dihydrothieno[2,3-d]pyrimidin-5-yl)methoxy)methyl)benzoic acid 99%
4-(((2-((3-Methoxybenzyl)carbamoyl)-4-oxo-1,4-dihydrothieno[2,3-d]pyrimidin-5-yl)methoxy)methyl)benzoic acid
Precaución: for Research Use Only


Formula: C₂₄H₂₁N₃O₆S
MDL Number: MFCD31558511
CAS Number: 869296-13-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty