Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-methoxy-N-(6-methoxy-3-methylbenzo[d]isoxazol-5-yl)benzenesulfonamide 99%

Supplier: AMBEED, INC
Warning

A1149116-10MG A1149116-1MG A1149116-25MG A1149116-50MG A1149116-5MG
76673-904EA 65 USD InStock
76673-904 76673-906 76673-908 76673-910 76673-912
5-Bromo-2-methoxy-N-(6-methoxy-3-methylbenzo[d]isoxazol-5-yl)benzenesulfonamide 99%
5-Bromo-2-methoxy-N-(6-methoxy-3-methylbenzo[d]isoxazol-5-yl)benzenesulfonamide
Precaución: for Research Use Only


Formula: C₁₆H₁₅BrN₂O₅S
MDL Number: MFCD31813647
CAS Number: 1832671-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty