Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1R,2R)-1,2-Bis[[[3,5-bis(tert-butyl)-2-hydroxyphenyl]methylene]amino]cyclohexane 98%

Supplier: AMBEED, INC
Warning

A1149287-25G A1149287-5G A1149287-100G
76673-946EA 61 USD InStock
76673-946 76673-948 77620-076
(1R,2R)-1,2-Bis[[[3,5-bis(tert-butyl)-2-hydroxyphenyl]methylene]amino]cyclohexane 98%
(1R,2R)-1,2-Bis[[[3,5-bis(tert-butyl)-2-hydroxyphenyl]methylene]amino]cyclohexane
Precaución: for Research Use Only


Formula: C₃₆H₅₄N₂O₂
MDL Number: MFCD00191800
CAS Number: 151433-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty