Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2,2-Tetrakis(4-methoxyphenyl)ethene 98%

Supplier: AMBEED, INC
Warning

A1149496-100MG A1149496-250MG A1149496-5G
76674-016EA 11 USD InStock
76674-016 76674-018 76674-020
1,1,2,2-Tetrakis(4-methoxyphenyl)ethene 98%
1,1,2,2-Tetrakis(4-methoxyphenyl)ethene
Precaución: for Research Use Only


Formula: C₃₀H₂₈O₄
MW: 452.5409 g/mol
MDL Number: MFCD00185484
CAS Number: 10019-24-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty