Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1,2,2-Tetrakis(4-iodophenyl)ethene 98%

Supplier: AMBEED, INC
Warning

A1149503-100MG A1149503-1G A1149503-250MG A1149503-5G
76674-022EA 19 USD InStock
76674-022 76674-024 76674-026 76674-028
1,1,2,2-Tetrakis(4-iodophenyl)ethene 98%
1,1,2,2-Tetrakis(4-iodophenyl)ethene
Precaución: for Research Use Only


Formula: C₂₆H₁₆I₄
MDL Number: MFCD31697671
CAS Number: 299914-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty