Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-4-(dibenzo[b,d]furan-3-yl)-6-phenyl-1,3,5-triazine 98%

Supplier: AMBEED, INC
Warning

A1149647-1G A1149647-250MG A1149647-5G
76674-110EA 32 USD InStock
76674-110 76674-112 76674-114
2-Chloro-4-(dibenzo[b,d]furan-3-yl)-6-phenyl-1,3,5-triazine 98%
2-Chloro-4-(dibenzo[b,d]furan-3-yl)-6-phenyl-1,3,5-triazine
Precaución: for Research Use Only


Formula: C₂₁H₁₂ClN₃O
MDL Number: MFCD32641703
CAS Number: 2142681-84-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty