Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-3-(4-Cyano-3-(3-((1-(2,3-dihydro-1H-inden-2-yl)-2-methylpropan-2-yl)amino)-2-hydroxypropoxy)phenyl)propanoic acid 97%

Supplier: AMBEED, INC
Warning

A1149674-1MG A1149674-5MG
76674-122EA 30 USD InStock
76674-122 76674-124
(R)-3-(4-Cyano-3-(3-((1-(2,3-dihydro-1H-inden-2-yl)-2-methylpropan-2-yl)amino)-2-hydroxypropoxy)phenyl)propanoic acid 97%
(R)-3-(4-Cyano-3-(3-((1-(2,3-dihydro-1H-inden-2-yl)-2-methylpropan-2-yl)amino)-2-hydroxypropoxy)phenyl)propanoic acid
Precaución: for Research Use Only


Formula: C₂₆H₃₂N₂O₄
MDL Number: MFCD19443794
CAS Number: 351490-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty