Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nickel(II) bromide 2-methoxyethyl ether complex 98%

Supplier: AMBEED, INC
Danger

A1149939-1G A1149939-5G A1149939-25G
76674-176EA 15 USD InStock
76674-176 76674-178 77620-100
Nickel(II) bromide 2-methoxyethyl ether complex 98%
Nickel(II) bromide 2-methoxyethyl ether complex
Precaución: for Research Use Only


Formula: C₆H₁₄Br₂NiO₃
MDL Number: MFCD00274339
CAS Number: 312696-09-6
UN: 1325
ADR: 4.1,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty