Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aluminium stearate 4-6% Al

Supplier: AMBEED, INC
Warning

A1150294-1000G A1150294-100G A1150294-500G
76674-230EA 19 USD InStock
76674-230 76674-232 76674-234
Aluminium stearate 4-6% Al
Aluminium stearate
Precaución: for Research Use Only


Formula: C₅₄H₁₀₅AlO₆
MDL Number: MFCD00036389
CAS Number: 637-12-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty